accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH3CNNNCH3CH3H3CH3CH3CHCl(?)

CAS RN 257639-98-8

IUPAC Name: N-butyl-N-ethyl-2,5-dimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride
Synonyms: N-butyl-N-ethyl-2,5-dimethyl-7-(2,4,6-trimethylphenyl)pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride
Molecular Weight (g/mol): 401
Molecular Formula: C23H33ClN4
InChi Key: ZWDLBNCJWNENFF-UHFFFAOYSA-N
SMILES: Cl.CCCCN(CC)C1=C2C(C)=CN(C2=NC(C)=N1)C1=C(C)C=C(C)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results