accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3H2CH3CCH2OOOOCH3CH2

CAS RN 25777-71-3

IUPAC Name: 2-[(2-methylprop-2-enoyl)oxy]ethyl 2-methylprop-2-enoate; methyl 2-methylprop-2-enoate
Synonyms: methyl methacrylate crosspolymer
Molecular Weight (g/mol): 298.34
Molecular Formula: C15H22O6
InChi Key: XBDAKYZJVVYINU-UHFFFAOYSA-N
SMILES: COC(=O)C(C)=C.CC(=C)C(=O)OCCOC(=O)C(C)=C
Quantity 
  • (1)
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)