accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNH2OONHSOOOSOOOSOOO

CAS RN 2580-78-1

IUPAC Name: disodium 1-amino-9,10-dioxo-4-({3-[2-(sulfonatooxy)ethanesulfonyl]phenyl}amino)-9,10-dihydroanthracene-2-sulfonate
Synonyms: remazol brilliant blue R
Molecular Weight (g/mol): 626.53
Molecular Formula: C22H16N2Na2O11S3
InChi Key: KUIXZSYWBHSYCN-UHFFFAOYSA-L
SMILES: [Na+].[Na+].NC1=C2C(=O)C3=CC=CC=C3C(=O)C2=C(NC2=CC=CC(=C2)S(=O)(=O)CCOS([O-])(=O)=O)C=C1S([O-])(=O)=O
Color 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)