Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Remazol Brilliant Blue R, pure

Catalog No. AC207550250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC207550250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC207550250 Supplier Thermo Scientific Chemicals Supplier No. 207550250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 2580-78-1 | C22H16N2Na2O11S3 | 626.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2580-78-1
Molecular Formula C22H16N2Na2O11S3
Molecular Weight (g/mol) 626.53
MDL Number MFCD00001215
InChI Key KUIXZSYWBHSYCN-UHFFFAOYSA-L
Synonym C.I. 61200
PubChem CID 17409
IUPAC Name disodium 1-amino-9,10-dioxo-4-({3-[2-(sulfonatooxy)ethanesulfonyl]phenyl}amino)-9,10-dihydroanthracene-2-sulfonate
SMILES [Na+].[Na+].NC1=C2C(=O)C3=CC=CC=C3C(=O)C2=C(NC2=CC=CC(=C2)S(=O)(=O)CCOS([O-])(=O)=O)C=C1S([O-])(=O)=O

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
UV Spectrometry Complies
Color Blue
Quantity 25 g
Physical Form Crystalline Powder
Formula Weight 626.55
Grade Extra Pure
Chemical Name or Material Remazol Brilliant Blue R

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.