accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOH2COHO

CAS RN 2583-25-7

IUPAC Name: 2-(prop-2-en-1-yl)propanedioic acid
Synonyms: 2-allyl-malonic acid
Molecular Weight (g/mol): 144.13
Molecular Formula: C6H8O4
InChi Key: ZDZVKPXKLLLOOA-UHFFFAOYSA-N
SMILES: OC(=O)C(CC=C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results