Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Allylmalonic acid

Catalog No. 0602310G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-023-10G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-023-10G Supplier Sigma Aldrich Supplier No. 0602310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,3-butadiene can be used:
  • In the synthesis of 1,6-hexanedial (adipic aldehyde) by bis-hydroformylation using rhodium as a catalyst
  • As a monomer for the synthesis of cis-1,4-polybutadiene by polymerization using cobalt(ii) and nickel(ii) complex catalysts
  • As a precursor for the selective synthesis of butene by hydrogenation reaction

Specifications

CAS 2583-25-7
Melting Point 102°C to 105°C
Molecular Formula C6H8O4
Linear Formula CH2=CHCH2CH(COOH)2
MDL Number MFCD00014347
Quantity 250 g
Molecular Weight (g/mol) 144.13
Percent Purity ≥98.0% (T)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.