accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 258864-54-9

IUPAC Name: trihexyl(tetradecyl)phosphanium chloride
Synonyms: trihexyl(tetradecyl)phosphanium chloride
Molecular Weight (g/mol): 519.32
Molecular Formula: C32H68ClP
InChi Key: JCQGIZYNVAZYOH-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCCCCCCCC[P+](CCCCCC)(CCCCCC)CCCCCC
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)