Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trihexyl(tetradecyl)phosphonium Chloride 97.0+%, TCI America™
SDP

Catalog No. T36235G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T36235G 5 g
T362325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T36235G Supplier TCI America Supplier No. T36235G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 258864-54-9
Molecular Formula C32H68ClP
Molecular Weight (g/mol) 519.32
MDL Number MFCD04112746
InChI Key JCQGIZYNVAZYOH-UHFFFAOYSA-M
Synonym Tetradecyl(trihexyl)phosphonium Chloride
PubChem CID 11375816
IUPAC Name trihexyl(tetradecyl)phosphanium;chloride
SMILES CCCCCCCCCCCCCC[P+](CCCCCC)(CCCCCC)CCCCCC.[Cl-]

Specifications

Color Colorless
Quantity 5 g
UN Number 1760
Formula Weight 519.32
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Trihexyl(tetradecyl)phosphonium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.