accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25954-13-6

IUPAC Name: ammonium ethyl carbamoylphosphonate
Synonyms: fosamine ammonium salt
Molecular Weight (g/mol): 170.11
Molecular Formula: C3H11N2O4P
InChi Key: OTSAMNSACVKIOJ-UHFFFAOYSA-N
SMILES: [NH4+].CCOP([O-])(=O)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results