accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3NNNOHCH3H3CH3C

CAS RN 25973-55-1

IUPAC Name: 2-(2H-1,2,3-benzotriazol-2-yl)-4,6-bis(2-methylbutan-2-yl)phenol
Synonyms: 2-(1,2,3-benzotriazol-2-yl)-4,6-bis(2-methylbutan-2-yl)phenol
Molecular Weight (g/mol): 351.49
Molecular Formula: C22H29N3O
InChi Key: ZMWRRFHBXARRRT-UHFFFAOYSA-N
SMILES: CCC(C)(C)C1=CC(N2N=C3C=CC=CC3=N2)=C(O)C(=C1)C(C)(C)CC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results