CAS RN 259793-96-9
IUPAC Name:
6-fluoro-3-oxo-3,4-dihydropyrazine-2-carboxamide
Synonyms:
favipiravir
Molecular Weight (g/mol):
157.1
Molecular Formula:
C5H4FN3O2
InChi Key:
ZCGNOVWYSGBHAU-UHFFFAOYSA-N
SMILES:
NC(=O)C1=NC(F)=CNC1=O
1
–
2
of
2
results
1
Promotions Available
Thermo Scientific Chemicals Favipiravir
Thermo Scientific Chemicals Favipiravir
CAS: 259793-96-9 Molecular Formula: C5H4FN3O2
2
R&D Systems™ Favipiravir
A Tocris product, part of R&D Systems. Viral RNA polymerase inhibitor