accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26027-38-3

IUPAC Name: 2-(4-nonylphenoxy)ethan-1-ol
Synonyms: nonoxynol amway all purpose spray adjuvant akyporox NP 150 polyoxyethylene (18) nonyl phenyl ether liponox ncy polyoxyethylene nonylphenyl ether nonoxinols patentex ovals ovals, patentex nonylphenoxypolyethoxyethanols
Molecular Weight (g/mol): 264.41
Molecular Formula: C17H28O2
InChi Key: KUXGUCNZFCVULO-UHFFFAOYSA-N
SMILES: CCCCCCCCCC1=CC=C(OCCO)C=C1
Quantity 
  • (1)
  • (4)
Physical Form 
  • (1)
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results