Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene Glycol Mono-4-nonylphenyl Ether (n=approx. 10), TCI America™
SDP

Catalog No. P070725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P070725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P070725G Supplier TCI America Supplier No. P070725G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 26027-38-3
MDL Number MFCD00148006

Specifications

Color Yellow
pH 6.7 (1% H2O soln. 25°C)
Quantity 25 g
Physical Form Liquid
Chemical Name or Material Polyethylene Glycol Mono-4-nonylphenyl Ether (n=approx. 10)
EINECSNumber (7)-0172
RTECSNumber RB1299090
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.