Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Igepal(R) CO-520, Spectrum™ Chemical
SDP

Catalog No. 18611014 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
20 kg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-014 20 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-611-014 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I115520KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 26027-38-3
Molecular Weight (g/mol) 440.63

Specifications

Color Colorless, Viscous
CAS Max % 1
Quantity 20 kg
Packaging Amber Glass Bottle
Refractive Index 1.493-1.497
Grade Ungraded
Chemical Name or Material Igepal CO to 520

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.