CAS RN 260970-57-8
IUPAC Name:
4-({[(tert-butoxy)carbonyl]amino}methyl)pyridine-2-carboxylic acid
Synonyms:
4-{[(tert-butoxycarbonyl)amino]methyl}pyridine-2-carboxylic acid
Molecular Weight (g/mol):
252.27
Molecular Formula:
C12H16N2O4
InChi Key:
BSNYTBOSHJFWQM-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NCC1=CC(=NC=C1)C(O)=O
1
–
1
of
1
results
1
4-[(tert-Butoxycarbonylamino)methyl]pyridine-2-carboxylic Acid 96.0+%, TCI America™
CAS: 260970-57-8 Molecular Formula: C12H16N2O4 Molecular Weight (g/mol): 252.27 MDL Number: MFCD02093452 InChI Key: BSNYTBOSHJFWQM-UHFFFAOYSA-N Synonym: 4-(Boc-aminomethyl)pyridine-2-carboxylic Acid