accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3O

CAS RN 26118-61-6

IUPAC Name: 10-(acetyloxy)decyl acetate
Synonyms: 10-(acetyloxy)decyl acetate
Molecular Weight (g/mol): 258.36
Molecular Formula: C14H26O4
InChi Key: NMOLZWLFORPIME-UHFFFAOYSA-N
SMILES: CC(=O)OCCCCCCCCCCOC(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)