accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOFOFFFOFFFFFFFFFFFFF

CAS RN 26131-32-8

IUPAC Name: methyl 2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propanoate
Synonyms: methyl 2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propanoate
Molecular Weight (g/mol): 510.1
Molecular Formula: C10H3F17O4
InChi Key: UMADKHFVSXRTGY-UHFFFAOYNA-N
SMILES: COC(=O)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results