accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CClH3CH3C

CAS RN 26189-59-3

IUPAC Name: (1-chloro-2-methylprop-1-en-1-yl)dimethylamine
Synonyms: (1-chloro-2-methylprop-1-en-1-yl)dimethylamine
Molecular Weight (g/mol): 133.62
Molecular Formula: C6H12ClN
InChi Key: GQIRIWDEZSKOCN-UHFFFAOYSA-N
SMILES: CN(C)C(Cl)=C(C)C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)