accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NH2ClNOO

CAS RN 26196-45-2

IUPAC Name: 2-chloro-5-nitrobenzene-1,4-diamine
Synonyms: 2-chloro-5-nitrobenzene-1,4-diamine
Molecular Weight (g/mol): 187.58
Molecular Formula: C6H6ClN3O2
InChi Key: VUNAQOGRLGNALG-UHFFFAOYSA-N
SMILES: NC1=CC(=C(N)C=C1Cl)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results