accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26227-73-6

IUPAC Name: N-(4-butylphenyl)-1-(4-methoxyphenyl)methanimine
Synonyms: MBBA
Molecular Weight (g/mol): 267.37
Molecular Formula: C18H21NO
InChi Key: FEIWNULTQYHCDN-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(C=C1)N=CC1=CC=C(OC)C=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results