Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Methoxybenzylidene)-4-butylaniline 98.0+%, TCI America™
SDP

Catalog No. M027525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M027525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M027525G Supplier TCI America Supplier No. M027525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 26227-73-6
Molecular Formula C18H21NO
Molecular Weight (g/mol) 267.37
MDL Number MFCD00009464
InChI Key FEIWNULTQYHCDN-UHFFFAOYSA-N
Synonym mbba, n-4-methoxybenzylidene-4-butylaniline, n-p-methoxybenzylidene-p-butylaniline, unii-s586t4jync, benzenamine, 4-butyl-n-4-methoxyphenyl methylene, 4-methoxybenzylidene-4'-n-butylaniline, ccris 4668, unii-gew7i7nn51, 4-butyl-n-4-methoxyphenyl methylene benzenamine, gew7i7nn51
PubChem CID 33363
IUPAC Name N-(4-butylphenyl)-1-(4-methoxyphenyl)methanimine
SMILES CCCCC1=CC=C(C=C1)N=CC1=CC=C(OC)C=C1

Specifications

Color White-Yellow
Boiling Point 176°C
Quantity 25 g
Formula Weight 267.37
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material N-(4-Methoxybenzylidene)-4-butylaniline
EINECSNumber (3)-1151&(3)-1212
RTECSNumber BW9550000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.