accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2623-82-7

H3C(S)BrOHO

CAS RN 2623-82-7

IUPAC Name: (2S)-2-bromooctanoic acid
Synonyms: (2S)-2-bromooctanoic acid
Molecular Weight (g/mol): 223.11
Molecular Formula: C8H15BrO2
InChi Key: GTGTXZRPJHDASG-ZETCQYMHSA-N
SMILES: CCCCCC[C@H](Br)C(O)=O
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)