CAS RN 2624332-04-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ BIIE 0246 hydrochloride
Potent, selective and competitive non-peptide NPY Y2 receptor antagonist; active in vivo