accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COH

CAS RN 26248-42-0

IUPAC Name: tridecan-1-ol
Synonyms: tridecanol
Molecular Weight (g/mol): 200.37
Molecular Formula: C13H28O
InChi Key: XFRVVPUIAFSTFO-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results