accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONHBr

CAS RN 2627-77-2

IUPAC Name: N-(4-bromophenyl)-2-hydroxybenzamide
Synonyms: 4'-BR salicylanilide
Molecular Weight (g/mol): 292.13
Molecular Formula: C13H10BrNO2
InChi Key: DKQMTUHJJPFJRL-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C(=O)NC1=CC=C(Br)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results