accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO(E)

CAS RN 26303-90-2

IUPAC Name: (4E)-dec-4-enoic acid
Synonyms: obtusilic acid
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: XKZKQTCECFWKBN-VOTSOKGWSA-N
SMILES: CCCCC\C=C\CCC(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results