accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONH2N

CAS RN 263396-44-7

IUPAC Name: 2-amino-3-(4-cyanophenyl)propanoic acid
Synonyms: 2-amino-3-(4-cyanophenyl)propanoic acid
Molecular Weight (g/mol): 190.2
Molecular Formula: C10H10N2O2
InChi Key: KWIPUXXIFQQMKN-UHFFFAOYNA-N
SMILES: NC(CC1=CC=C(C=C1)C#N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)