CAS RN 2634129-70-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Janelia Fluor™ 646b, Maleimide
Blinking fluorescent dye (Reactive group: maleimide). Application: super resolution microscopy (SRM) including SMLM