accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNOOHOH

CAS RN 263717-53-9

IUPAC Name: 4-[1,5-bis(4-hydroxyphenyl)-4-propyl-2,3-dihydro-1H-pyrazol-3-ylidene]cyclohexa-2,5-dien-1-one
Synonyms: propylpyrazoletriol
Molecular Weight (g/mol): 386.45
Molecular Formula: C24H22N2O3
InChi Key: UOSWGERPQQOSHS-UHFFFAOYSA-N
SMILES: CCCC1=C(N(NC1=C1C=CC(=O)C=C1)C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)