accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH3CON

CAS RN 26391-06-0

IUPAC Name: 2-cyano-N,N-diethylacetamide
Synonyms: N,N-diethylcyanoacetamide N,N-diethylaminocyanoacetamide
Molecular Weight (g/mol): 140.19
Molecular Formula: C7H12N2O
InChi Key: RYSHIRFTLKZVIH-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)CC#N
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results