accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26392-49-4

HClH3CCH3NH2

CAS RN 26392-49-4

IUPAC Name: hydrogen 3-(aminomethyl)heptane chloride
Synonyms: hydrogen 2-ethylhexylamine chloride
Molecular Weight (g/mol): 165.71
Molecular Formula: C8H20ClN
InChi Key: JNWRGTDFPPYRAD-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CCCCC(CC)CN
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results