accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2640-58-6

IUPAC Name: 5-amino-2-{[(benzyloxy)carbonyl]amino}pentanoic acid
Synonyms: 5-amino-2-{[(benzyloxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 266.3
Molecular Formula: C13H18N2O4
InChi Key: ZYGRWJVRLNJIMR-UHFFFAOYNA-N
SMILES: NCCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)