accessibility menu, dialog, popup

UserName

Viewing profile as user:
HONHOHNONOOCl

CAS RN 264218-23-7

IUPAC Name: 3-[(3-chloro-4-hydroxyphenyl)amino]-4-(2-nitrophenyl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 3-[(3-chloro-4-hydroxyphenyl)amino]-4-(2-nitrophenyl)-1H-pyrrole-2,5-dione
Molecular Weight (g/mol): 359.72
Molecular Formula: C16H10ClN3O5
InChi Key: PQCXVIPXISBFPN-UHFFFAOYSA-N
SMILES: OC1=CC=C(NC2=C(C(=O)NC2=O)C2=CC=CC=C2[N+]([O-])=O)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)