accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaCH3HSOO

CAS RN 26473-47-2

IUPAC Name: sodium 2-methyl-3-sulfanylpropanoate
Synonyms: sodium 2-methyl-3-sulfanylpropanoate
Molecular Weight (g/mol): 142.15
Molecular Formula: C4H7NaO2S
InChi Key: FOTPRJUNQGWLCM-UHFFFAOYNA-M
SMILES: [Na+].CC(CS)C([O-])=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results