accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26533-34-6

IUPAC Name: 2-methyloctan-3-ol
Synonyms: 2-methyloctan-3-ol
Molecular Weight (g/mol): 144.26
Molecular Formula: C9H20O
InChi Key: DIVBBSLQUDHECU-UHFFFAOYNA-N
SMILES: CCCCCC(O)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results