CAS RN 265980-25-4
IUPAC Name:
N-(2,3-diphenyl-2,5-dihydro-1,2,4-thiadiazol-5-ylidene)methanamine hydrobromide
Synonyms:
N-(2,3-diphenyl-1,2,4-thiadiazol-5-ylidene)methanamine hydrobromide
Molecular Weight (g/mol):
348.26
Molecular Formula:
C15H14BrN3S
InChi Key:
YJYGOWVFDGULLL-UHFFFAOYSA-N
SMILES:
Br.CN=C1SN(C(=N1)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
R&D Systems™ SCH 202676 hydrobromide
CAS: 265980-25-4 Molecular Formula: C15H14BrN3S