accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26629-33-4

CH3OOH3CHNOCH3

CAS RN 26629-33-4

IUPAC Name: methyl 2-acetamidopropanoate
Synonyms: alanine-N-acetyl,methyl ester
Molecular Weight (g/mol): 145.16
Molecular Formula: C6H11NO3
InChi Key: FQGVVDYNRHNTCK-UHFFFAOYNA-N
SMILES: COC(=O)C(C)NC(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)