CAS RN 2665674-77-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Alkyne-cRGD
Integrin ligand for degrader development and targeted delivery, with alkyne click handle for conjugation