accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOFFFOFFFOFFFOFFFFFFFFFFFFFFFFFFF

CAS RN 26738-51-2

IUPAC Name: 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-icosafluoro-5,8,11-tris(trifluoromethyl)-3,6,9,12-tetraoxapentadecane
Synonyms: 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-icosafluoro-5,8,11-tris(trifluoromethyl)-3,6,9,12-tetraoxapentadecane
Molecular Weight (g/mol): 784.11
Molecular Formula: C14HF29O4
InChi Key: NOCLWIPRQIWFMR-UHFFFAOYNA-N
SMILES: FC(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F
Boiling Point 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)