Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-Icosafluoro-5,8,11-tris(trifluoromethyl)-3,6,9,12-tetraoxapentadecane 92+%, TCI America™
SDP

Catalog No. I104425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I104425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. I104425G Supplier TCI America Supplier No. I104425G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 26738-51-2
Molecular Formula C14HF29O4
Molecular Weight (g/mol) 784.11
MDL Number MFCD00054717
InChI Key NOCLWIPRQIWFMR-UHFFFAOYNA-N
Synonym 2h-perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxatetradecane, 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-icosafluoro-5,8,11-tris trifluoromethyl-3,6,9,12-tetraoxapentadecane, 3,6,9,12-tetraoxapentadecane, 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-eicosafluoro-5,8,11-tris trifluoromethyl, freon e 4, freon e-4, 2h-perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxapentadecane, 1,1,1,2,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy-3-1,2,2,2-tetrafluoroethoxy propane
PubChem CID 117327
IUPAC Name 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-icosafluoro-5,8,11-tris(trifluoromethyl)-3,6,9,12-tetraoxapentadecane
SMILES FC(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F

Specifications

Color Colorless
Boiling Point 194°C
Quantity 25 g
Formula Weight 784.11
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-Icosafluoro-5,8,11-tris(trifluoromethyl)-3,6,9,12-
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.