accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2683-57-0

IUPAC Name: 5-(diethylamino)pentan-1-ol
Synonyms: 5-(diethylamino)pentan-1-ol
Molecular Weight (g/mol): 159.27
Molecular Formula: C9H21NO
InChi Key: RUQDFMATAGUGMU-UHFFFAOYSA-N
SMILES: CCN(CC)CCCCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results