CAS RN 26833-87-4
IUPAC Name:
(2R,3S,6R)-4-methoxy-16,18-dioxa-10-azapentacyclo[11.7.0.0²,⁶.0⁶,¹⁰.0¹⁵,¹⁹]icosa-1(20),4,13,15(19)-tetraen-3-yl 1-methyl 3-hydroxy-3-(4-hydroxy-4-methylpentyl)butanedioate
Synonyms:
omacetaxine mepesuccinate
Molecular Weight (g/mol):
545.63
Molecular Formula:
C29H39NO9
InChi Key:
HYFHYPWGAURHIV-ZEDNPHJLSA-N
SMILES:
COC(=O)CC(O)(CCCC(C)(C)O)C(=O)O[C@H]1[C@@H]2C3=CC4=C(OCO4)C=C3CCN3CCC[C@]23C=C1OC
1
–
1
of
1
results
1
Tocris Bioscience™ Homoharringtonine
CAS: 26833-87-4 Molecular Formula: C29H39NO9