accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNO

CAS RN 2687-94-7

IUPAC Name: 1-octylpyrrolidin-2-one
Synonyms: caprylyl pyrrolidone
Molecular Weight (g/mol): 197.32
Molecular Formula: C12H23NO
InChi Key: WPPOGHDFAVQKLN-UHFFFAOYSA-N
SMILES: CCCCCCCCN1CCCC1=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)