accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOH

CAS RN 26896-48-0

IUPAC Name: [8-(hydroxymethyl)tricyclo[5.2.1.0²,⁶]decan-3-yl]methanol
Synonyms: tricyclodecanedimethanol tricyclodecane-4,8-dimethanol
Molecular Weight (g/mol): 196.29
Molecular Formula: C12H20O2
InChi Key: OTLDLKLSNZMTTA-UHFFFAOYNA-N
SMILES: OCC1CCC2C3CC(CC3CO)C12
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)