CAS RN 27012-22-2
IUPAC Name:
5-methyl-2-phenylpyridine
Synonyms:
5-methyl-2-phenylpyridine
Molecular Weight (g/mol):
169.23
Molecular Formula:
C12H11N
InChi Key:
ZYLPQYYLLRBVOK-UHFFFAOYSA-N
SMILES:
CC1=CC=C(N=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
5-Methyl-2-phenylpyridine 98.0+%, TCI America™
CAS: 27012-22-2 Molecular Formula: C12H11N Molecular Weight (g/mol): 169.227 MDL Number: MFCD04114180 InChI Key: ZYLPQYYLLRBVOK-UHFFFAOYSA-N Synonym: 6-Phenyl-3-picoline PubChem CID: 47467 IUPAC Name: 5-methyl-2-phenylpyridine SMILES: CC1=CN=C(C=C1)C2=CC=CC=C2