CAS RN 27025-41-8
L-Glutathione, oxidized
CAS: 27025-41-8 Molecular Formula: C20H32N6O12S2 Molecular Weight (g/mol): 612.63 MDL Number: MFCD00150701 InChI Key: YPZRWBKMTBYPTK-IOKZUGQQNA-N Synonym: oxiglutatione,glutathione disulfide,gssg,oxidized glutathione,glutathione disulphide,glutathione-ssg,l-glutathione oxidized,oxigluthione,glutathone disulfide,glutathione oxidized PubChem CID: 65359 ChEBI: CHEBI:17858 IUPAC Name: (2S)-2-amino-5-[[(2R)-3-[[(2R)-2-[[(4S)-4-amino-4-carboxybutanoyl]amino]-3-(carboxymethylamino)-3-oxopropyl]disulfanyl]-1-(carboxymethylamino)-1-oxopropan-2-yl]amino]-5-oxopentanoic acid SMILES: N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O
L(-)-Glutathione, oxidized, 98%
CAS: 27025-41-8 Molecular Formula: C20H32N6O12S2 Molecular Weight (g/mol): 612.63 MDL Number: MFCD00150701 InChI Key: YPZRWBKMTBYPTK-IOKZUGQQNA-N Synonym: oxiglutatione,glutathione disulfide,gssg,oxidized glutathione,glutathione disulphide,glutathione-ssg,l-glutathione oxidized,oxigluthione,glutathone disulfide,glutathione oxidized PubChem CID: 65359 ChEBI: CHEBI:17858 IUPAC Name: 2-amino-4-[(2-{[2-(4-amino-4-carboxybutanamido)-2-[(carboxymethyl)carbamoyl]ethyl]disulfanyl}-1-[(carboxymethyl)carbamoyl]ethyl)carbamoyl]butanoic acid SMILES: N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O
Glutathione Oxidized, MP Biomedicals™
CAS: 27025-41-8 Molecular Formula: C20H32N6O12S2 Molecular Weight (g/mol): 612.63 MDL Number: MFCD00150701 InChI Key: YPZRWBKMTBYPTK-IOKZUGQQNA-N Synonym: oxiglutatione,glutathione disulfide,gssg,oxidized glutathione,glutathione disulphide,glutathione-ssg,l-glutathione oxidized,oxigluthione,glutathone disulfide,glutathione oxidized PubChem CID: 65359 ChEBI: CHEBI:17858 IUPAC Name: (2S)-2-amino-4-{[(1R)-2-{[(2R)-2-[(4S)-4-amino-4-carboxybutanamido]-2-[(carboxymethyl)carbamoyl]ethyl]disulfanyl}-1-[(carboxymethyl)carbamoyl]ethyl]carbamoyl}butanoic acid SMILES: N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O
Glutahione oxidized, For HPLC analysis, 98%, MP Biomedicals™
CAS: 27025-41-8 Molecular Formula: C20H32N6O12S2 Molecular Weight (g/mol): 612.63 MDL Number: MFCD00150701 InChI Key: YPZRWBKMTBYPTK-IOKZUGQQNA-N Synonym: oxiglutatione,glutathione disulfide,gssg,oxidized glutathione,glutathione disulphide,glutathione-ssg,l-glutathione oxidized,oxigluthione,glutathone disulfide,glutathione oxidized PubChem CID: 65359 ChEBI: CHEBI:17858 SMILES: N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O
L(-)-Glutathione, oxidized hydrate, 95%
CAS: 27025-41-8 Molecular Formula: C20H32N6O12S2 Molecular Weight (g/mol): 612.63 MDL Number: MFCD00150701 InChI Key: YPZRWBKMTBYPTK-IOKZUGQQNA-N Synonym: oxiglutatione,glutathione disulfide,gssg,oxidized glutathione,glutathione disulphide,glutathione-ssg,l-glutathione oxidized,oxigluthione,glutathone disulfide,glutathione oxidized PubChem CID: 65359 ChEBI: CHEBI:17858 IUPAC Name: (2S)-2-amino-5-[[(2R)-3-[[(2R)-2-[[(4S)-4-amino-4-carboxybutanoyl]amino]-3-(carboxymethylamino)-3-oxopropyl]disulfanyl]-1-(carboxymethylamino)-1-oxopropan-2-yl]amino]-5-oxopentanoic acid SMILES: N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O
Glutathione oxidized form 98.0+%, TCI America™
CAS: 27025-41-8 Molecular Formula: C20H32N6O12S2 Molecular Weight (g/mol): 612.63 MDL Number: MFCD00150701 InChI Key: YPZRWBKMTBYPTK-IOKZUGQQNA-N Synonym: oxiglutatione,glutathione disulfide,gssg,oxidized glutathione,glutathione disulphide,glutathione-ssg,l-glutathione oxidized,oxigluthione,glutathone disulfide,glutathione oxidized PubChem CID: 65359 ChEBI: CHEBI:17858 IUPAC Name: (2S)-2-amino-4-{[(1R)-2-{[(2R)-2-[(4S)-4-amino-4-carboxybutanamido]-2-[(carboxymethyl)carbamoyl]ethyl]disulfanyl}-1-[(carboxymethyl)carbamoyl]ethyl]carbamoyl}butanoic acid SMILES: N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O