Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione Oxidized, MP Biomedicals™

Catalog No. MP021511935 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021511935 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021511935 Supplier MP Biomedicals, Inc Supplier No. 02151193.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protects against oxidative damage. Glutathione Oxidized, MP Biomedicals is the oxidised dimeric form of glutathione (GSH), a naturally occuring tripeptide that takes part in the body's detoxification mechanisms.

Acts as a hydrogen acceptor in enzymic determination of NADP and NADPH.

  • In vivo, Glutathione disulfide (GSSG) is reduced back to glutathione (GSH) via the NADPH-dependent enzyme glutathione reductase
  • The ratio of GSH to GSSG is often used to gauge the level of oxidative stress in cells, with higher concentrations of GSSG implying more oxidative stress
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27025-41-8
Molecular Formula C20H32N6O12S2
Molecular Weight (g/mol) 612.63
MDL Number MFCD00150701
InChI Key YPZRWBKMTBYPTK-IOKZUGQQNA-N
Synonym oxiglutatione, glutathione disulfide, gssg, oxidized glutathione, glutathione disulphide, glutathione-ssg, l-glutathione oxidized, oxigluthione, glutathone disulfide, glutathione oxidized
PubChem CID 65359
ChEBI CHEBI:17858
IUPAC Name (2S)-2-amino-4-{[(1R)-2-{[(2R)-2-[(4S)-4-amino-4-carboxybutanamido]-2-[(carboxymethyl)carbamoyl]ethyl]disulfanyl}-1-[(carboxymethyl)carbamoyl]ethyl]carbamoyl}butanoic acid
SMILES N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O

Specifications

Melting Point 178°C (decomposition)(literature)
pH 1 to 6
Quantity 500 mg
Solubility Information Soluble in water (3.3%): Clear, Colorless Solution.
Optical Rotation (c=1, water) –85 to –110°
Formula Weight 612.64
Percent Purity ≥98%
Grade Not Available
Physical Form Crystalline Powder
Recommended Storage +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.