accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27060-75-9

IUPAC Name: 1-bromo-4-methoxy-2-methylbenzene
Synonyms: 1-bromo-4-methoxy-2-methylbenzene
Molecular Weight (g/mol): 201.06
Molecular Formula: C8H9BrO
InChi Key: BLZNSXFQRKVFRP-UHFFFAOYSA-N
SMILES: COC1=CC=C(Br)C(C)=C1
Boiling Point 
  • (4)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (4)
  • (1)
  • (2)
  • (2)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)