Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-methoxytoluene, 97%, Thermo Scientific™

Catalog No. AC294031000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC294031000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC294031000 Supplier Thermo Scientific Chemicals Supplier No. 294031000

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 27060-75-9
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD00060687
InChI Key BLZNSXFQRKVFRP-UHFFFAOYSA-N
Synonym 2-bromo-5-methoxytoluene, 4-bromo-3-methylanisole, benzene, 1-bromo-4-methoxy-2-methyl, 4-bromo-3-methyl-anisole, unii-okr329132j, 1-bromo-2-methyl-4-methoxybenzene, 1-brom-4-methoxy-2-methylbenzol, pubchem3796, 3-methyl-4-bromoanisole, acmc-1cg4z
PubChem CID 117915
IUPAC Name 1-bromo-4-methoxy-2-methylbenzene
SMILES CC1=C(C=CC(=C1)OC)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 111.0°C to 112.0°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.5600 to 1.5620
Quantity 100 g
Specific Gravity 0.91
Formula Weight 201.06
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-methoxytoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.