accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOOOCH3

CAS RN 27215-22-1

IUPAC Name: 1-(6-methylheptyl) 2-(4-methylphenyl) benzene-1,2-dicarboxylate
Synonyms: 1-(6-methylheptyl) 2-(4-methylphenyl) phthalate
Molecular Weight (g/mol): 368.47
Molecular Formula: C23H28O4
InChi Key: OJTKBTGQAKWTED-UHFFFAOYSA-N
SMILES: CC(C)CCCCCOC(=O)C1=CC=CC=C1C(=O)OC1=CC=C(C)C=C1
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results